4-(2-Methylpropoxy)benzonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 4-iso-Butoxybenzonitrile

OR900341-1G OR900341-5G
APOSOR900341-1GEA 40.6 EUR
APOSOR900341-1G APOSOR900341-5G
4-(2-Methylpropoxy)benzonitrile 98%
4-(2-Methylpropoxy)benzonitrile
Formel: C₁₁H₁₃NO
Molecular Weight: 175.227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274895
CAS-Nummer: 5203-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR