4-(Cyclopropylmethoxy)benzonitrile 98%

Lieferant: Apollo Scientific

OR900345-1G OR900345-25G OR900345-5G
APOSOR900345-1GEA 145 EUR
APOSOR900345-1G APOSOR900345-25G APOSOR900345-5G
4-(Cyclopropylmethoxy)benzonitrile 98%
4-(Cyclopropylmethoxy)benzonitrile
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11136694
CAS-Nummer: 1019619-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR