4-(Diethylamino)benzonitril 98%

Lieferant: Apollo Scientific

OR900360-100G OR900360-25G OR900360-5G
APOSOR900360-100GEA 647 EUR
APOSOR900360-100G APOSOR900360-25G APOSOR900360-5G
4-(Diethylamino)benzonitril 98%
4-(Diethylamino)benzonitril
Formel: C₁₁H₁₄N₂
Molecular Weight: 174.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019754
CAS-Nummer: 2873-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR