1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione 98%

Lieferant: Apollo Scientific

OR900371-5G
APOSOR900371-5GEA 527 EUR
APOSOR900371-5G
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione 98%
1-(2-Hydroxyphenyl)pyrrolidine-2,5-dione
Formel: C₁₀H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00461771
CAS-Nummer: 79252-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR