1,3-Dicyclopentylurea 95%

Lieferant: Apollo Scientific

OR900409-1G
APOSOR900409-1GEA 364 EUR
APOSOR900409-1G
1,3-Dicyclopentylurea 95%
1,3-Dicyclopentylurea
Formel: C₁₁H₂₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13258925
CAS-Nummer: 58713-33-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR