1-Bromo-3-(cyclopropylsulphonyl)benzene ≥98%

Lieferant: Apollo Scientific

OR900454-250MG
APOSOR900454-250MGEA 159 EUR
APOSOR900454-250MG
1-Bromo-3-(cyclopropylsulphonyl)benzene ≥98%
1-Bromo-3-(cyclopropylsulphonyl)benzene
Formel: C₉H₉BrO₂S
Molecular Weight: 261.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21879801
CAS-Nummer: 19433-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR