3-(Cyclopropylmethoxy)propanenitrile 97%

Lieferant: Apollo Scientific

OR900474-1G
APOSOR900474-1GEA 270 EUR
APOSOR900474-1G
3-(Cyclopropylmethoxy)propanenitrile 97%
3-(Cyclopropylmethoxy)propanenitrile
Formel: C₇H₁₁NO
Molecular Weight: 125,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11136705
CAS-Nummer: 1019568-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR