4-Chlor-2-methylbenzylalkohol 98%

Lieferant: Apollo Scientific

OR900545-5G
APOSOR900545-5GEA 166 EUR
APOSOR900545-5G
4-Chlor-2-methylbenzylalkohol 98%
4-Chlor-2-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06201140
CAS-Nummer: 129716-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR