1-Bromo-4-(ethenyloxy)benzene 98%

Lieferant: Apollo Scientific

Synonyme: 1-Bromo-4-(vinyloxy)benzene

OR900581-1G
APOSOR900581-1GEA 364 EUR
APOSOR900581-1G
1-Bromo-4-(ethenyloxy)benzene 98%
1-Bromo-4-(ethenyloxy)benzene
Formel: C₈H₇BrO
Molecular Weight: 199.045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21755355
CAS-Nummer: 1005-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR