Ethyl 2-cyano-3-methyl-butanoate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-cyano-3-methylbutanoate

OR900594-1G
APOSOR900594-1GEA 502 EUR
APOSOR900594-1G
Ethyl 2-cyano-3-methyl-butanoate 97%
Ethyl 2-cyano-3-methyl-butanoate
Formel: C₈H₁₃NO₂
Molecular Weight: 155,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258612
CAS-Nummer: 3213-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR