4-(Pyrrolidinocarbonyl)phenol 98%
Lieferant: Apollo Scientific
OR900668-1G
OR900668-5G
APOSOR900668-1GEA
328
EUR
APOSOR900668-1G
APOSOR900668-5G
4-(Pyrrolidinocarbonyl)phenol 98%
4-(Pyrrolidinocarbonyl)phenol
Formel:
C₁₁H₁₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07633880 CAS-Nummer: 478929-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...