4-Methoxy-m-toluidin 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-3-methylphenylamine, 4-Methoxy-3-methylaniline

OR900687-25G
APOSOR900687-25GEA 79.9 EUR
APOSOR900687-25G
4-Methoxy-m-toluidin 95%
4-Methoxy-m-toluidin
Formel: C₈H₁₁NO
Molecular Weight: 137.1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779234
CAS-Nummer: 136-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR