N-(2,2-Dimethylpropyl)-2,2-dimethylpropanamide 98%
Lieferant: Apollo Scientific
OR900774-1G
APOSOR900774-1GEA
400
EUR
APOSOR900774-1G
N-(2,2-Dimethylpropyl)-2,2-dimethylpropanamide 98%
N-(2,2-Dimethylpropyl)-2,2-dimethylpropanamide
Formel:
C₁₀H₂₁NO Molecular Weight: 171,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00509282 CAS-Nummer: 20826-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...