4-Methoxy-3-methylbenzamide 98%

Lieferant: Apollo Scientific

OR900803-1G
APOSOR900803-1GEA 806 EUR
APOSOR900803-1G
4-Methoxy-3-methylbenzamide 98%
4-Methoxy-3-methylbenzamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09600240
CAS-Nummer: 932123-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR