(6-Brom-3-pyridinyl)methanol 98%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol

OR900843-25G
APOSOR900843-25GEA 108 EUR
APOSOR900843-25G
(6-Brom-3-pyridinyl)methanol 98%
(6-Brom-3-pyridinyl)methanol
Formel: C₆H₆BrNO
Molecular Weight: 188.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974508
CAS-Nummer: 122306-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR