Methyl 5-formylpyrrole-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-formyl-1H-pyrrole-2-carboxylate

OR900846-5G
APOSOR900846-5GEA 105 EUR
APOSOR900846-5G
Methyl 5-formylpyrrole-2-carboxylate 98%
Methyl 5-formylpyrrole-2-carboxylate
Formel: C₇H₇NO₃
Molecular Weight: 153.135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878545
CAS-Nummer: 1197-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR