3,4-Diethylpyrrole 98%
Lieferant: Apollo Scientific
Synonyme:
3,4-Diethyl-1H-pyrrole
OR900851-1G
APOSOR900851-1GEA
478
EUR
APOSOR900851-1G
3,4-Diethylpyrrole 98%
3,4-Diethylpyrrole
Formel:
C₈H₁₃N Molecular Weight: 123,196 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062571 CAS-Nummer: 16200-52-5 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...