(4-Carbamoyl-2-methylphenyl)boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

OR900945-1G OR900945-250MG
APOSOR900945-1GEA 638 EUR
APOSOR900945-1G APOSOR900945-250MG
(4-Carbamoyl-2-methylphenyl)boronic acid, pinacol ester 97%
(4-Carbamoyl-2-methylphenyl)boronic acid, pinacol ester
Formel: C₁₄H₂₀BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18732747
CAS-Nummer: 631911-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR