1-Methyl-4-imidazolsulfonylchlorid 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9010-25G
APOSOR9010-25GEA
0
EUR
APOSOR9010-25G
1-Methyl-4-imidazolsulfonylchlorid 95+%
1-Methyl-4-imidazolsulfonylchlorid
Formel:
C₄H₅ClN₂O₂S Schmelzpunkt: 85…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068060 CAS-Nummer: 137049-00-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...