4-Aminobenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Aminophenyl)methanol, 4-(Hydroxymethyl)anilin

OR9011-100G OR9011-25G OR9011-5G
APOSOR9011-100GEA 108 EUR
APOSOR9011-100G APOSOR9011-25G APOSOR9011-5G
4-Aminobenzylalkohol 98%
4-Aminobenzylalkohol
Formel: C₇H₉NO
Molecular Weight: 123.15 g/mol
Schmelzpunkt: 61…63 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00014782
CAS-Nummer: 623-04-1
EINECS: 210-767-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR