4-Aminobenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Aminophenyl)methanol, 4-(Hydroxymethyl)anilin
OR9011-100G
OR9011-25G
OR9011-5G
APOSOR9011-100GEA
108
EUR
APOSOR9011-100G
APOSOR9011-25G
APOSOR9011-5G
4-Aminobenzylalkohol 98%
4-Aminobenzylalkohol
Formel:
C₇H₉NO Molecular Weight: 123.15 g/mol Schmelzpunkt: 61…63 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00014782 CAS-Nummer: 623-04-1 EINECS: 210-767-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...