2-(4-Aminophenyl)oxazole 98%

Lieferant: Apollo Scientific

OR901543-1G OR901543-5G
APOSOR901543-1GEA 0 EUR
APOSOR901543-1G APOSOR901543-5G
2-(4-Aminophenyl)oxazole 98%
2-(4-Aminophenyl)oxazole
Formel: C₉H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11040381
CAS-Nummer: 62882-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR