2-(4-Aminophenyl)oxazole 98%
Lieferant: Apollo Scientific
OR901543-1G
OR901543-5G
APOSOR901543-1GEA
0
EUR
APOSOR901543-1G
APOSOR901543-5G
2-(4-Aminophenyl)oxazole 98%
2-(4-Aminophenyl)oxazole
Formel:
C₉H₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040381 CAS-Nummer: 62882-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...