2-Amino-4-methylphenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Amino-4-methylphenylboronic acid, pinacol ester

OR901564-1G
APOSOR901564-1GEA 168 EUR
APOSOR901564-1G
2-Amino-4-methylphenylboronsäurepinakolester 97%
2-Amino-4-methylphenylboronsäurepinakolester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233,12 g/mol
Schmelzpunkt: 40…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06795679
CAS-Nummer: 863578-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR