1,5-Naphthyridin-2(1H)-one 98%

Lieferant: Apollo Scientific

OR901584-1G
APOSOR901584-1GEA 59.6 EUR
APOSOR901584-1G
1,5-Naphthyridin-2(1H)-one 98%
1,5-Naphthyridin-2(1H)-one
Formel: C₈H₆N₂O
Molecular Weight: 146.146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11877925
CAS-Nummer: 10261-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR