Methyl-3-methoxypicolinat 95%
Lieferant: Apollo Scientific
OR901627-1G
APOSOR901627-1GEA
151
EUR
APOSOR901627-1G
Methyl-3-methoxypicolinat 95%
Methyl-3-methoxypicolinat
Formel:
C₈H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06412569 CAS-Nummer: 24059-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...