5-Bromo-N-methylpyridin-2-amine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-(N-methylamino)pyridine, 5-Bromo-2-methylaminopyridine

OR901635-5G
APOSOR901635-5GEA 58 EUR
APOSOR901635-5G
5-Bromo-N-methylpyridin-2-amine 98%
5-Bromo-N-methylpyridin-2-amine
Formel: C₆H₇BrN₂
Molecular Weight: 187.0400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272096
CAS-Nummer: 84539-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR