8-Methyl-2-chinolinol 98%

Lieferant: Apollo Scientific

OR901652-1G OR901652-5G
APOSOR901652-1GEA 49.3 EUR
APOSOR901652-1G APOSOR901652-5G
8-Methyl-2-chinolinol 98%
8-Methyl-2-chinolinol
Formel: C₁₀H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13195790
CAS-Nummer: 4053-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR