8-Chloroquinolin-2-ol 98%
Lieferant: Apollo Scientific
OR901653-100G
OR901653-25G
APOSOR901653-100GEA
328
EUR
APOSOR901653-100G
APOSOR901653-25G
8-Chloroquinolin-2-ol 98%
8-Chloroquinolin-2-ol
Formel:
C₉H₆ClNO Molecular Weight: 179.603 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427166 CAS-Nummer: 23981-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...