2-Chlor-8-methylchinolin 98%

Lieferant: Apollo Scientific

OR901660-5G
APOSOR901660-5GEA 134 EUR
APOSOR901660-5G
2-Chlor-8-methylchinolin 98%
2-Chloro-8-methylquinoline
Formel: C₁₀H₈ClN
Molecular Weight: 177.6300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11108682
CAS-Nummer: 4225-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR