2-Amino-5-chloro-3-methoxypyridine 96%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-3-methoxypyridin-2-amine

OR901750-1G
APOSOR901750-1GEA 306 EUR
APOSOR901750-1G
2-Amino-5-chloro-3-methoxypyridine 96%
2-Amino-5-chloro-3-methoxypyridine
Formel: C₆H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD17015845
CAS-Nummer: 1242336-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR