6,7,8,9-Tetrahydro-5H-imidazo-[1,2-a]azepine-3-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR9018-1G
APOSOR9018-1GEA 497 EUR
APOSOR9018-1G
6,7,8,9-Tetrahydro-5H-imidazo-[1,2-a]azepine-3-sulphonyl chloride 95%
6,7,8,9-Tetrahydro-5H-imidazo-[1,2-a]azepine-3-sulphonyl chloride
Formel: C₈H₁₁ClN₂O₂S
Molecular Weight: 234,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08235265
CAS-Nummer: 914637-93-7
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR