6-Chloro-3-methoxypyridazin-4-amine 98%

Lieferant: Apollo Scientific

OR901840-1G OR901840-250MG
APOSOR901840-1GEA 751 EUR
APOSOR901840-1G APOSOR901840-250MG
6-Chloro-3-methoxypyridazin-4-amine 98%
6-Chloro-3-methoxypyridazin-4-amine
Formel: C₅H₆ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00234095
CAS-Nummer: 14369-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR