2-(3-Bromopyridin-2-yl)acetonitrile 98%
Lieferant: Apollo Scientific
OR901848-1G
APOSOR901848-1GEA
97.3
EUR
APOSOR901848-1G
2-(3-Bromopyridin-2-yl)acetonitrile 98%
2-(3-Bromopyridin-2-yl)acetonitrile
Formel:
C₇H₅BrN₂ Molecular Weight: 197,032 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999977 CAS-Nummer: 122851-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...