6-Chloro-8-methylquinoline 96%

Lieferant: Apollo Scientific

OR901870-10G OR901870-1G OR901870-5G
APOSOR901870-10GEA 0 EUR
APOSOR901870-10G APOSOR901870-1G APOSOR901870-5G
6-Chloro-8-methylquinoline 96%
6-Chloro-8-methylquinoline
Formel: C₁₀H₈ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD10686879
CAS-Nummer: 19655-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR