6-Nitro-1H-quinolin-2-one 98%

Lieferant: Apollo Scientific

OR901876-1G
APOSOR901876-1GEA 132 EUR
APOSOR901876-1G
6-Nitro-1H-quinolin-2-one 98%
6-Nitro-1H-quinolin-2-one
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11501896
CAS-Nummer: 64495-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR