6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR9019-1G
APOSOR9019-1GEA 749 EUR
APOSOR9019-1G
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-sulphonyl chloride 95%
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole-3-sulphonyl chloride
Formel: C₆H₇ClN₂O₂S
Lagertemperatur: Gefrierschrank
MDL: MFCD08235266
CAS-Nummer: 914637-94-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR