2-Hydroxy-5-methoxypyridine 98%

Lieferant: Apollo Scientific

OR901930-1G
APOSOR901930-1GEA 539 EUR
APOSOR901930-1G
2-Hydroxy-5-methoxypyridine 98%
2-Hydroxy-5-methoxypyridine
Formel: C₆H₇NO₂
Schmelzpunkt: 102…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11044353
CAS-Nummer: 61941-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR