3-Amino-5-chloro-2-methoxypyridine 98%

Lieferant: Apollo Scientific

OR901932-1G
APOSOR901932-1GEA 342 EUR
APOSOR901932-1G
3-Amino-5-chloro-2-methoxypyridine 98%
3-Amino-5-chloro-2-methoxypyridine
Formel: C₆H₇ClN₂O
Molecular Weight: 158,586 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07375045
CAS-Nummer: 886373-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR