Ethyl-6-chlorpicolinat 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate

OR901965-25G
APOSOR901965-25GEA 82.8 EUR
APOSOR901965-25G
Ethyl-6-chlorpicolinat 97%
Ethyl-6-chlorpicolinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878438
CAS-Nummer: 21190-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR