2-(Ethylamino)-3-nitro-5-picolin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Ethylamino-5-methyl-3-nitropyridine

OR902004-100G OR902004-25G OR902004-5G
APOSOR902004-100GEA 647 EUR
APOSOR902004-100G APOSOR902004-25G APOSOR902004-5G
2-(Ethylamino)-3-nitro-5-picolin 98%
2-(Ethylamino)-3-nitro-5-picolin
Formel: C₈H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09862688
CAS-Nummer: 106690-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR