5,6,7,8-Tetrahydroquinolin-2(1H)-one 97%

Lieferant: Apollo Scientific

OR902091-1G
APOSOR902091-1GEA 243 EUR
APOSOR902091-1G
5,6,7,8-Tetrahydroquinolin-2(1H)-one 97%
5,6,7,8-Tetrahydroquinolin-2(1H)-one
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00456287
CAS-Nummer: 54802-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR