2-Chlor-3-methyl-4-nitropyridin-N-oxid 96%

Lieferant: Apollo Scientific

OR902101-1G
APOSOR902101-1GEA 290 EUR
APOSOR902101-1G
2-Chlor-3-methyl-4-nitropyridin-N-oxid 96%
2-Chlor-3-methyl-4-nitropyridin-N-oxid
Formel: C₆H₅ClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01194031
CAS-Nummer: 60323-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR