2-Acetamido-3-methylpyridine 96%

Lieferant: Apollo Scientific

Synonyme: 2-Acetamido-3-picoline, N-(3-Methylpyridin-2-yl)acetamide

OR902134-5G
APOSOR902134-5GEA 156 EUR
APOSOR902134-5G
2-Acetamido-3-methylpyridine 96%
2-Acetamido-3-methylpyridine
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00495347
CAS-Nummer: 7463-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR