1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95%

Lieferant: Apollo Scientific

OR902251-1G OR902251-250MG
APOSOR902251-1GEA 281 EUR
APOSOR902251-1G APOSOR902251-250MG
1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95%
1-(Pyrrolidin-2-ylmethyl)pyrrolidine
Formel: C₉H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06653330
CAS-Nummer: 195311-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR