Benzo[d]oxazol-2-ylmethanol 95%

Lieferant: Apollo Scientific

OR902294-1G OR902294-5G
APOSOR902294-1GEA 0 EUR
APOSOR902294-1G APOSOR902294-5G
Benzo[d]oxazol-2-ylmethanol 95%
Benzo[d]oxazol-2-ylmethanol
Formel: C₈H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07368333
CAS-Nummer: 77186-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR