3-Methoxy-1-n,1-N-dimethylbenzene-1,4-diamine 95%
Lieferant: Apollo Scientific
Synonyme:
3-Methoxy-1-N,1-N-dimethylbenzene-1,4-diamine
OR902314-1G
APOSOR902314-1GEA
48
EUR
APOSOR902314-1G
3-Methoxy-1-n,1-N-dimethylbenzene-1,4-diamine 95%
3-Methoxy-1-n,1-N-dimethylbenzene-1,4-diamine
Formel:
C₉H₁₄N₂O Molecular Weight: 166,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16249531 CAS-Nummer: 7474-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...