3-(2-Methoxyethoxy)benzonitril 95%

Lieferant: Apollo Scientific

OR902326-1G
APOSOR902326-1GEA 114 EUR
APOSOR902326-1G
3-(2-Methoxyethoxy)benzonitril 95%
3-(2-Methoxyethoxy)benzonitril
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,1998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907640
CAS-Nummer: 80407-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR