Methyl 4-amino-5-methylthiophene-2-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR902339-1G
OR902339-5G
APOSOR902339-1GEA
151
EUR
APOSOR902339-1G
APOSOR902339-5G
Methyl 4-amino-5-methylthiophene-2-carboxylate 97%
Methyl 4-amino-5-methylthiophene-2-carboxylate
Formel:
C₇H₉NO₂S Schmelzpunkt: 91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14694722 CAS-Nummer: 501082-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...