3(5)-Isopropyl-4-nitro-1H-pyrazole 98%
Lieferant: Apollo Scientific
Synonyme:
3-Isopropyl-4-nitro-1H-pyrazole
OR902381-1G
APOSOR902381-1GEA
207
EUR
APOSOR902381-1G
3(5)-Isopropyl-4-nitro-1H-pyrazole 98%
3(5)-Isopropyl-4-nitro-1H-pyrazole
Formel:
C₆H₉N₃O₂ Molecular Weight: 155,155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14657963 CAS-Nummer: 51355-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...